Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Color of Varnish (Gardner Color) Discs

$822.00

Specifications

Certifications/Compliance ASTM D-1544, AOCS Td 1a
Type Color of Varnish l Disc
For Use With (Equipment) Varnish comparator kits 705-V2, 705-VA2 or 705-VA4
View More Specs

Products 2
Catalog Number Mfr. No. Range Price Quantity & Availability  
Catalog Number Mfr. No. Range Price Quantity & Availability  
09-043-434
View Documents
Lovibond™
243030
1 to 9
Each for $822.00
Add to Cart
 
09-053-435
View Documents
Lovibond™
243040
9 to 18
Each for $822.00
Add to Cart
 
Description

Description

Specifications

Specifications

ASTM D-1544, AOCS Td 1a
Color of Varnish l Disc
Varnish comparator kits 705-V2, 705-VA2 or 705-VA4
SDS
Documents

Documents

Product Certifications
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.