Learn More
Lovibond™ Color of Water Comparator Disc for 711 Nesslerizer Comparator
Manufacturer:  Lovibond™ 284150
Catalog No. 09-054-140
Add to cart
Description
Description
Color of Water Comparator Disc for 711 Nesslerizer Comparator
Specifications
Specifications
0 to 30 Color Units
Color of Water Comparator Disc
Pt-Co or APHA
Measuring the color of potable water in terms of an easily reproducible color standard
711 Nesslerizer comparator