Learn More
Lovibond™ Colorimeter Reagent Set: Ozone
For use with MC500, MD600 colorimeter and SP600 spectrophotometer.
Manufacturer:  Lovibond™ 517791BT
Catalog No. 09-534-608
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Includes: 100 each of DPD No. 1 tablets, DPD No. 3 tablets and glycine tablets
Description
Description
For use with MC500, MD600 colorimeter and SP600 spectrophotometer.
Specifications
Specifications
Ozone Colorimeter Reagent Set
MC500, MD600 colorimeter and SP600 spectrophotometer
100 each of DPD No. 1 tablets, DPD No. 3 tablets and glycine tablets