Learn More
Lovibond™ Free Ammonia Reagent Set
Manufacturer:  Lovibond™ 531800
 View more versions of this product
Catalog No. 09-534-453
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Free Ammonia Reagent Solution
0.01 to 0.5mg/L
Analysis of free ammonia
13.1
0.13 oz.
Potassium Hydroxide <10%, Sodium Hypochlorite solution 0.5 to 3%
Liquid
Free ammonia SC450 and MC500 colorimeters, SP600 spectrophotometer
20°C
4mL