Lovibond™ MC500 Colorimeter Replacement Reagents: Chlorine Available on GSA/VA Contract for Federal Government customers only.
For use with MC500 multi-parameter colorimeter
Manufacturer: Lovibond™ 517711BT
 View all options for this product
Catalog No. 15-398-482
  Check Availability
Add to Cart
    Product Type Chlorine Reagent Set
    Range 0.01 to 6mg/L
    For Use With (Application) Water quality testing
    For Use With (Equipment) MC500 multi-parameter colorimeter
    Includes DPD No. 1 tablets (100 pack), DPD No. 3 tablets (100 pack)
    No. of Tests 100.0
    Method DPD
    • Parameter: Chlorine
    • Method: DPD
  • Description & Specifications
Related Products
Product Content Feedback