Lovibond™ MC500, MD600 Colorimeter Replacement Reagents: pH value

For use with MC500 and MD600 multi-parameter colorimeter

Manufacturer: Lovibond™ 511770BT

 View more versions of this product

Catalog No. 09-534-469

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Water quality testing
    For Use With (Equipment) MC500, MD600, SC450, MD100 pool control line, aquaPRO3 and aquaPRO6 photometers and scuba II electronic pool tester
    No. of Tests 100.0
    pH Range 5.1
    Storage Requirements 20°C
    Shelf Life 10 Years
    Method Phenol red
    Color Orange
    Product Type Phenol Red Photometer Tablet
    Format Tablet
    Composition Sodium Chloride 90 to 100%, non-hazardous components <2%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit