Lovibond™ Model 611-A Aqua Tester Interchangeable Color Discs
Optically standardized; guaranteed never to fade or change color
Manufacturer: Lovibond™ 611-100/PRC
 View all options for this product
Catalog No. 15-398-212
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Range 0 to 500 color units
    Includes Necessary glassware, reagent
    For Use With (Equipment) 611-A Aqua Tester
    Product Type Interchangeable Color Disc
    Nine or 10 precision standards permanently mounted in plastic disc. Discs are supplied with all the necessary glassware and reagents.
  • Description & Specifications
Related Products
Product Content Feedback