Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Optical Glass Colorimetry Cells

Made from optical glass to the highest standards.

Manufacturer:  Lovibond™ 605960

 View more versions of this product

Catalog No. 09-053-344


Add to cart

Description

Description

Precision fused cells in a range of dimensions and path lengths. Note: For instruments equipped with a heater unit and whenever cells are subjected to thermal shock, it is recommended that borosilicate cells be used.

  • Series: W600 Optical
  • 21 x 40mm cells
  • Use only with nonheated samples (up to 40°C)

Specifications

Specifications

Model Fs, PFXi-195, 880, 950 and 995 instruments with non-heated samples (up to 40°C)
Optical Glass
10mm
Colorimetry Cell
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit