Lovibond™ Pocket Comparators Chlorine Color Discs for Orthotolidine Method
Manufacturer: Lovibond™ 332D/PRC
 View more versions of this product
Catalog No. 15-398-165
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance ISO 7393-2
    For Use With Comparator 705
    Methods Acid O-Tolidine
    Parameter Chlorine
    Range 0 to 1mg/L
    Reagents Ortho-Tolidine solution
    Standards 0, 0.05, 0.1, 0.2, 0.3, 0.4, 0.5, 0.6, 0.8, 1.0ppm
    Type Chlorine color disc
  • Description & Specifications
Related Products
Product Content Feedback