Learn More
Lovibond™ Pocket Comparators DPD Tablets
For use with DPD Color Discs and Pocket Comparators
Manufacturer:  Lovibond™ 433 IT
 View more versions of this product
Catalog No. 15-398-102C
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
DPD Color Discs and Pocket comparators
DPD No. 3 Tablets
0.2 to 4mg/L
10 Tablets/Strip
Combined Chlorine