Lovibond™ Pocket Comparators Low-Range Chlorine Color Discs for DPD Procedure
Manufacturer: Lovibond™ 430 D/PRC
 View more versions of this product
Catalog No. 15-398-101A
 N/A
N/A
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance ISO 7393-2
    For Use With Comparator 705
    For Use With (Application) Tests free, combined and total chlorine
    Includes Color disc, reagents, labware and directions
    Methods DPD
    Parameter Chlorine
    Range 0.2 to 4mg/L
    Reagents DPD No. 1 and DPD No. 3 Tablets
    Standards 0.2, 0.4, 0.6, 0.8, 1.0, 1.5, 2.0, 2.5, 3.0, 4.0ppm
    Type Chlorine color disc
  • Description & Specifications
Related Products
Product Content Feedback