Learn More
Lovibond™ Powder Packs™ for Free Chlorine
Premeasured unit dose DPD reagents for water quality testing
Manufacturer:  Lovibond™ 530103
 View more versions of this product
Catalog No. 09-053-219
Add to cart
Description
Description
  • For use in SC450™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed foil packets eliminate risk of cross-contamination
  • Available for 5, 10, and 25mL sample sizes
  • Extended shelf life of up to 5 years
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted DPD method
Specifications
Specifications
US EPA
SC450 colorimeter and Aqua Comparator™ test kits, Hach™ instruments and test kits, spectrophotometer
1000 Pack
0.33 oz.
DPD Free Chlorine Reagent
Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
Powder
0 to 2mg/L
10mL
Safety and Handling
Safety and Handling
ShelfLife : 5 Years