Lovibond™ Powder Packs™: Silica HR Molybdate
Pre-measured, unit dose reagents for water quality testing
Manufacturer: Lovibond™ 531723
 View all options for this product
Catalog No. 09-053-752
  Check Availability
Add to Cart
    Product Type HR Molybdate
    Range 0 to 100mg/L
    For Use With (Application) Water quality testing
    For Use With (Equipment) Colorimeter
    • Simply tear open Powder Pack™ and pour reagent into sample
    • Large design makes them easier to handle
    • “Easy Open” notch makes it simple to tear open the package
    • Easily create a “V”, to allow user to pour reagent into sample
    • Beveled edge prevents reagent from getting stuck in the corners
  • Description & Specifications
Related Products
Product Content Feedback