Learn More
Alfa Aesar™ Low Form Inconel Crucible
Manufacturer:  Alfa Aesar™ 35994KT
 View more versions of this product
Catalog No. AA35994KT
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
55mL
43mm
Inconel
Top: 47mm; Bottom: 38mm
Low Form
Inconel Crucible