Learn More
United Scientific™ Low Retention Filter Micropipette Tips, Racked
SDPSCI_ED
These Racked Low Retention Micropipette Tips are disposable tips intended for use with most brands of pipettors.
Supplier:  United Scientific™ UNP11065
 View more versions of this product
Catalog No. S8538020
Add to Cart
Description
Description
  • Manufactured from 100% pure porous polyethylene, filters are designed to prevent liquids and aerosols from entering the body of the micropipette to protect against cross-contamination.
  • Deliver 10x lower retention levels with viscous samples, as compared to standard pipette tips.
  • Best suited for biological applications, these tips are non-cytotoxic, non-hemolytic, and have a 99.9% bacterial filtration efficiency (BFE).
  • Tips are manufactured from ultra pure USP Class VI certified, virgin polypropylene.
  • Free of DNase, RNase and pyrogen.
Specifications
Specifications
Pipette Filter Tips 2 to 20 μL
Filtered
Natural
Autoclavable
USP Class VI Certified
Steam
Rack, 10 x 96
10 Racks of 96
Single-use
Yes
20 μL
Sterile
White
UNP11065
Pipetting
Universal
Low Retention
960 Pk.
Universal Fit Micro
Polyethylene