Learn More
Fisherbrand™ Low-Retention Pipet Tips, Nonfilter
Increase accuracy by reducing sample retention
$69.80
Specifications
Model Low Retention
Color Natural
View More Specs
Products 1
Catalog Number Description Volume (Metric) Tip Style Price Quantity & Availability  
Catalog Number Description Volume (Metric) Tip Style Price Quantity & Availability  
02-717-155
View Documents
Low RetentionNon-sterile, Rack 1000 μL Extended Length Pack of 1000 for $69.80
Case of 5 PK for $293.50
Add to cart
Description
Description
Increase accuracy by reducing sample retention
  • Low retention surface minimizes samples loss
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Natural color
Specifications
Specifications
Low Retention
Natural