Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Low-Retention Pipet Tips - Filtered

Catalog No. 02717166 Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
10 μL
1000 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
02-717-166 1000 μL
02-717-158 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-717-166

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Increase accuracy by reducing sample retention

  • Low retention surface minimizes samples loss
  • Certified by independent laboratory
  • Nonsterile: RNase, DNase, DNA, and Pyrogen Free; Sterile: RNase, DNase, DNA, Bioburden, PCR Inhibitor, and Pyrogen Free
  • Natural color

TRUSTED_SUSTAINABILITY

Specifications

Description Low retention, Filtered, Sterile, Rack
Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Natural
Model Low retention
Format Rack
Tip Style Low Retention
For Use With (Equipment) Universal Fit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.