Learn More
Malvern Panalytical Low Volume Flow Cell Gasket Component x2
Supplier:  Malvern Panalytical 10047186
Encompass
Low Volume Flow cell Gasket Component for NanoSight NS300 and NS500 (non-zeta) instruments (Pack of 2 engraved NTA0066 - Low Volume Flow Cell Gasket component)
Catalog No. NC1691727
Explore available promotions