Learn More
LowDensityLRP Mouse anti-Human, Janelia Fluor 646, Clone: A2Mr alpha-2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10064808JF646
Catalog No. NB064808646
Add to cart
Description
Description
LowDensityLRP Monoclonal antibody specifically detects LowDensityLRP in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
LowDensityLRP
A2Mr alpha-2
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
LRP1
Purified alpha2 macroglobulin receptor
Protein G purified
Store at 4°C in the dark.
Monoclonal
Recognises an epitope in the extracellular region of the 500kD alpha chain.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
A2MRTGFBR5, Alpha-2-macroglobulin receptorMGC88725, APOER, Apolipoprotein E receptor, APR, CD91, CD91 antigen, EC 1.1.1.94, EC 3.4.21.9, FLJ16451, IGFBP3R, low density lipoprotein receptor-related protein 1, LRP, LRP-1, prolow-density lipoprotein receptor-related protein 1, TbetaR-V/LRP-1/IGFBP-3 receptor, type V tgf-beta receptor
Mouse
IgG1
0.1mL
Primary
4035
Human