Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOX Mouse anti-Human, Clone: 9A11, Abnova™

Mouse monoclonal antibody raised against partial recombinant human LOX.

Manufacturer:  Abnova Corporation MAB12485

Catalog No. 89-934-488


Add to cart

Description

Description

The protein encoded by this gene is an extracellular copper enzyme that initiates the crosslinking of collagens and elastin. The enzyme catalyzes oxidative deamination of the epsilon-amino group in certain lysine and hydroxylysine residues of collagens and lysine residues of elastin. In addition to crosslinking extracellular matrix proteins, the encoded protein may have a role in tumor suppression. [provided by RefSeq]

Specifications

Specifications

LOX
9A11
Mouse monoclonal antibody raised against partial recombinant human LOX.
In PBS, pH 7.3 (1% BSA, 50% glycerol, 0.02% sodium azide)
MGC105112
Mouse
IgG1
50 uL
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (1:100) Immunofluorescence (1:50-1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Liquid
LOX
Recombinant protein corresponding to amino acids 22-168 of human LOX.
Protein A/G purification
RUO
Primary
4015
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit