Learn More
Alfa Aesar™ Loxistatin
Manufacturer:  Alfa Aesar™ J65822LB0
 View more versions of this product
Catalog No. AAJ65822LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
5mg
-30°C to -10°C
Loxistatin
88321-09-9
342.43
Soluble in DMSO, DMF or ethanol
White
Powder
A cell-permeable, irreversible inhibitor of cysteine proteases
C17H30N2O5
EST; E-64d