Learn More
Aviva Systems Biology Corp LOXL1 Antibody - N-terminal region (ARP54780_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54780_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOXL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LOXL1. Alternate names: LOL, LOXLApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61311