Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOXL2 Mouse anti-Human, Clone: OTI8B2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526873

Catalog No. PIMA526873


Add to cart

Description

Description

Lysyl Oxidase (LO) is an extracellular, copper-dependent enzyme that initiates the cross-linking of collagens and elastin by catalyzing the oxidative deamination of peptidyl lysine to alpha-aminoadipic-delta-semialdehyde. Members of LO family have diverse functions which include tumor suppression and cell adhesion and senescence. LOXL2, a 87KDa protein, is a novel member of this family. It contains 3 potential N-linked glycosylation sites, 4 scavenger receptor cysteine-rich (SRCR) domains and highly conserved copper-binding and Lysyl-tyrosylquinone cofactor sites. It is a component of extracellular matrix that might be involved in cell adhesion and senescence. Role of LOXL2 has been implicated in maligt transformation.
Specifications

Specifications

LOXL2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y4K0
LOR2, WS9-14, lysyl oxidase homolog 2, lysyl oxidase related 2, lysyl oxidase-like protein 2, lysyl oxidase-related protein 2, lysyl oxidase-related protein WS9-14
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4017
Immunohistochemistry (Paraffin)
OTI8B2
Unconjugated
LOXL2
Liquid
LOXL2
Human recombit protein fragment corresponding to amino acids 152-450 of LOXL2 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit