Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LOXL3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548462

Catalog No. PA548462


Add to cart

Description

Description

Predicted to react with mouse based on sequence homology.

LOXL3 is a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
Specifications

Specifications

LOXL3
Polyclonal
PBS with 0.09% sodium azide
P58215
Lysyl oxidase homolog 3, 143-, Lysyl oxidase-like protein 3, LOXL3, LOXL
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84695
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LOXL3
Liquid
LOXL3
KLH conjugated synthetic peptide between 715-744 amino acids from the C-terminal region of human LOXL3.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit