Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ LP 922056

Notum inhibitor; activates Wnt signaling

$173.30 - $730.00

Specifications

CAS 1365060-22-5
Chemical Name or Material 2-[(6-Chloro-7-cyclopropylthieno[3,2-d]pyrimidin-4-yl)thio]acetic acid
Molecular Formula C11H9ClN2O2S2
Purity 98%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
66-911-0
View Documents
Tocris Bioscience™
6691/10
10 mg
Each for $173.30
Add to cart
 
66-915-0
View Documents
Tocris Bioscience™
6691/50
50 mg
Each for $730.00
Add to cart
 
Description

Description

  • Notum pectinacetylesterase inhibitor.
  • Activates wnt signalling in vitro (EC50 values are 21 nM in human and 55 nM in mouse cell-based assay).
  • Increases cortical bone thickness in mouse model of bone growth.
Specifications

Specifications

1365060-22-5
2-[(6-Chloro-7-cyclopropylthieno[3,2-d]pyrimidin-4-yl)thio]acetic acid
C11H9ClN2O2S2
98%
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit