Learn More
Thermo Scientific™ LP Vortex Mixer; 100-240V, 50/60Hz, US plug PROMO
Choose to continuously mix or pulse samples
Manufacturer:  Thermo Scientific™ 88880017F5
Catalog No. 11-676-331F5
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
0 to 3,000rpm
8.3 in.
83 mm
154 mm
6.8 lb.
100 to 240V
CE
USA
Mixers
100 to 240V 50/60Hz
3.3 in.
6.1 in.
210 mm
0.5 kg (1.1 lb.)
3.1 kg
50/60Hz
Continuous / Touch
2 Years
8.3 x 6.1 x 3.3 in. (210 x 154 x 83 mm)