Learn More
Thermo Scientific™ LP Vortex Mixer Available on GSA/VA Contract for Federal Government customers only.
Choose to continuously mix or pulse samples with our low profile vortex mixer.
Manufacturer:  Thermo Scientific™ 88880018
 View more versions of this product
Catalog No. 11-676-332
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Add to cart
Description
Description
Choose to continuously mix or pulse samples with the Thermo Scientific™ LP Vortex Mixer.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0 to 3,000rpm
8.3 in.
83mm
154mm
6.8 lb.
100 to 240V
CE
Cord set w/various plugs
Mixers
100 to 240V 50/60Hz
3.3 in.
6.1 in.
210mm
0.5kg (1.1 lb.)
3.1kg
50/60Hz
Continuous / Touch
2 Years
8.3 x 6.1 x 3.3 in. (210 x 154 x 83mm)
Documents
Accessories
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.