Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPA receptor 1 Polyclonal Antibody, Huabio
Rabbit Polyclonal Antibody
Supplier: HUABIO RESEARCH INC 09025


Description
LPA receptor 1 Polyclonal antibody specifically detects LPA receptor 1 in Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
LPA receptor 1 | |
Polyclonal | |
Unconjugated | |
LPAR1 | |
AI326300; Edg2; edg-2; endothelial differentiation lysophosphatidic acid G-protein-coupled receptor 2; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 2; Gpcr26; Gpcr91; GPR26; G-protein coupled receptor 26; Kdt2; LPA receptor 1; LPA1; LPA-1; LPAR1; Lysophosphatidic acid receptor 1; lysophosphatidic acid receptor Edg-2; Mrec1.3; rec.1.3; rec1.3; ventricular zone gene 1; Vzg1; vzg-1 | |
Synthetic peptide within N-terminal Mouse LPA receptor 1. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.2% BSA, 25% glycerol and 0.05% sodium azide; pH 7.4 | |
P61793 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14745 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction