Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPAM-1 Rat anti-Mouse, BV421, Clone: DATK32 , BD Horizon™
Rat Monoclonal Antibody
Supplier: BD Biosciences 566294
Description
LPAM-1 Monoclonal antibody specifically detects LPAM-1 in Mouse samples. It is validated for Flow cytometry.Specifications
LPAM-1 | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Rat | |
25 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
DATK32 | |
Brilliant Violet 421 | |
Itga4Itgb7; α4β7 Integrin; alpha 4 beta 7 integrin; CD49dβ7 | |
AKR/Cum mouse T lymphoma line TK1 | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed. | |
IgG2a κ, CDF IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?
Provide Content Correction