Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPAR1 Mouse anti-Human, Clone: OTI1G6, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503737S

Catalog No. 50-167-4556


Add to cart

Description

Description

EDG2 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids There are eight known members of the EDG receptor family and they are implicated in mediating growth-related effects such as induction of cellular proliferation, alterations in differentiation and survival, and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis.
Specifications

Specifications

LPAR1
Monoclonal
0.8 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q92633
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
Mouse
IgG2b
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1902
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1G6
Unconjugated
LPAR1
Liquid
LPAR1
Full length human recombit protein of human LPAR1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit