Learn More
LPAR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 221651AP
Catalog No. 50-173-3753
Add to cart
Description
Description
EDG4 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids. There are eight known members of the EDG receptor family and they are implicated in mediating growth-related effects such as induction of cellular proliferation, alterations in differentiation and survival, and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis. EDG receptors are developmentally regulated and differ in tissue distribution. They couple to multiple types of G proteins to signal through ras and MAP kinase, rho, phospholipase C, and several protein tyrosine kinases. EDG4 is expressed in testes, ovarian tumor, and leukocyte containing tissues.
Specifications
Specifications
LPAR4
0.4 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BLG2, Q99677
G protein coupled receptor 23, GPR23, LPA 4, LPA receptor 4, LPA4, LPAR4, P2RY9, P2Y purinoceptor 9, P2Y5 LIKE, P2Y5 like receptor, P2Y9, Purinergic receptor 9
Rabbit
IgG
150 μL
-20°C
Primary
2846, 302378, 78134
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LPAR4
Liquid
LPAR4
LPAR4 Fusion Protein Ag17817
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat