Learn More
Aviva Systems Biology Corp LPCAT1 Antibody - middle region (ARP49884_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP49884_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LPCAT1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LPCAT1. Alternate names: AYTL2, FLJ12443, PFAAP3, lpcatApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-60208