Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LPCAT1 Polyclonal Antibody
LPCAT1 Polyclonal Antibody
Supplier: Thermo Scientific PA5106278
Description
LPCAT1 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
LPCAT1 | |
Polyclonal | |
Unconjugated | |
LPCAT1 | |
1-acylglycerophosphocholine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; 2900035H07Rik; acetyl-CoA:lyso-PAF acetyltransferase; Acetyl-CoA:lyso-platelet-activating factor acetyltransferase; acyl-CoA:lysophosphatidylcholin; acyl-CoA:lysophosphatidylcholine acyltransferase 1; acyltransferase like 2; acyltransferase-like 2; AGPAT10; AGPAT9; Aytl2; BB137372; BC005662; C87117; LPC acyltransferase 1; LPCAT; LPCAT1; LPCAT-1; lyso-PAF acetyltransferase; lysoPAFAT; lysoPC acyltransferase 1; lysophosphatidylcholine acyltransferase 1; mLPCAT1; PFAAP3; Phosphonoformate immuno-associated protein 3; rd11; regulated by phosphonoformate; RGD1311599 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
210992, 361467, 79888 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q1HAQ0, Q3TFD2, Q8NF37 | |
LPCAT1 | |
A synthesized peptide derived from human LPCAT1(Accession Q8NF37), corresponding to amino acid residues F174-A224. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction