Learn More
LPCAT4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 179051AP
Catalog No. 50-173-1865
Add to cart
Description
Description
Members of the 1-acylglycerol-3-phosphate O-acyltransferase (EC 2.3.1.51) family, such as AGPAT7, catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA), a precursor in the biosynthesis of all glycerolipids. Both LPA and PA are involved in signal transduction.
Specifications
Specifications
LPCAT4
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q643R3, Q6NVG1
1 AGP acyltransferase 7, 1 AGPAT 7, Acyltransferase like 3, AGPAT7, AYTL3, LPAAT eta, LPCAT4, LPEAT2
Rabbit
IgG
150 μL
-20°C
Primary
254531, 296048, 99010
Western Blot
Unconjugated
LPCAT4
Liquid
Lpcat4
LPCAT4 Fusion Protein Ag12281
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat