Learn More
LPIN1 Mouse anti-Human, Clone: OTI7F5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806141
Catalog No. 50-166-7704
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene represents a candidate gene for human lipodystrophy, characterized by loss of body fat, fatty liver, hypertriglyceridemia, and insulin resistance. Mouse studies suggest that this gene functions during normal adipose tissue development and may also play a role in human triglyceride metabolism.
Specifications
Specifications
LPIN1
Monoclonal
Unconjugated
LPIN1
Lyophilized
LPIN1
Full length human recombit protein of human LPIN1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI7F5
PBS with 8% trehalose and no preservative; pH 7.3
Q14693
PAP1
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
23175