Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LPIN1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA116997

Catalog No. PIPA116997


Only null left
Add to Cart

Description

Description

Suggested positive control: 3T3-L1 cell lysates, antigen standard for LPIN1 (transient overexpression lysate).

The Lipin family of nuclear proteins contains three members: Lipin-1, Lipin-2 and Lipin-3, all of which contain a nuclear signal sequence, a highly conserved amino-terminal (NLIP) domain and a carboxy-terminal (CLIP) domain. LPIN1 (Lipin-1) is crucial for normal adipose tissue development and metabolism. LPIN1 selectively activates a subset of PGC-1A; target pathways, including fatty acid oxidation and mitochondrial oxidative phosphorylation by inducing expression of the nuclear receptor PPARA. LPIN1 also inactivates the lipogenic program and suppresses circulating lipid levels. An abundance of LPIN1 promotes fat accumulation and insulin sensitivity, whereas a deficiency in LPIN1 may deter normal adipose tissue development, resulting in insulin resistance and lipodystrophy, a heterogeneous group of disorders characterized by loss of body fat, fatty liver, hypertriglyceridemia and insulin resistance.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LPIN1
Polyclonal
Unconjugated
LPIN1
4631420P06; DKFZp781P1796; Fatty liver dystrophy protein; Fld; KIAA0188; lipin 1; Lipin1; lipin-1; LPIN1; mKIAA0188; PAP1; Phosphatidate phosphatase LPIN1
Rabbit
Antigen affinity chromatography
RUO
14245, 23175, 313977
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.8 mg/mL
PBS with 30% glycerol and 0.1% sodium azide
Q14693, Q91ZP3
LPIN1
A synthetic peptide to an internal region (within residues 300-400) of the human LPIN1 protein.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.