Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPIN2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549612

Catalog No. PA549612

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Plays important roles in controlling the metabolism of fatty acids at differents levels. Acts as a magnesium-dependent phosphatidate phosphatase enzyme which catalyzes the conversion of phosphatidic acid to diacylglycerol during triglyceride, phosphatidylcholine and phosphatidylethanolamine biosynthesis in the reticulum endoplasmic membrane. Acts also as a nuclear transcriptional coactivator for PPARGC1A to modulate lipid metabolism.
Specifications

Specifications

LPIN2
Polyclonal
PBS with 0.09% sodium azide
Q92539
Phosphatidate phosphatase LPIN2, Lipin-2, LPIN2, KIAA0249
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9663
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LPIN2
Liquid
LPIN2
KLH conjugated synthetic peptide between 262-288 amino acids from the Central region of human LPIN2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit