Learn More
LPIN3 Mouse anti-Human, Clone: OTI3H3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF804945
Catalog No. 50-166-7227
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Humans lipodystrophy is characterized by loss of body fat, fatty liver, hypertriglyceridemia, and insulin resistance. Mice carrying mutations in the fatty liver dystrophy gene have similar phenotypes. Through positional cloning, the mouse gene responsible for fatty liver dystrophy was isolated and designated Lpin1. The nuclear protein encoded by Lpin1 was named lipin. Lpin1 mRNA was expressed at high levels in adipose tissue and was induced during differentiation of preadipocytes. These results indicated that lipin is required for normal adipose tissue development and provided a candidate gene for human lipodystrophy. Through database searches, mouse and human EST and genomic sequences with similarities to Lpin1 were identified. These included two related mouse genes and three human homologs. Human LPIN1 gene has been mapped to 2p25.; linkages of fat mass and serum leptin levels to this same region have been noted. Human LPIN2 and LPIN3 mapped to chromosomes 18p11 and 20q11-q12, respectively. The mouse genes encoding Lpin1, Lpin2, and Lpin3 mapped to chromosome 12, 17, and 2, respectively.
Specifications
Specifications
LPIN3
Monoclonal
Unconjugated
LPIN3
Lyophilized
LPIN3
Full length human recombit protein of human LPIN3 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI3H3
PBS with 8% trehalose and no preservative; pH 7.3
Q9BQK8
LIPN3L, SMP2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
64900