Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LPL Mouse anti-Human, Clone: OTI4G2, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503810

Catalog No. 50-167-4611


Add to cart

Description

Description

LPL encodes lipoprotein lipase, which is expressed in heart, muscle, and adipose tissue. LPL functions as a homodimer, and has the dual functions of triglyceride hydrolase and ligand/bridging factor for receptor-mediated lipoprotein uptake. Severe mutations that cause LPL deficiency result in type I hyperlipoproteinemia, while less extreme mutations in LPL are linked to many disorders of lipoprotein metabolism.
Specifications

Specifications

LPL
Monoclonal
0.53 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P06858
HDLCQ11, LIPD
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4023
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI4G2
Unconjugated
LPL
Liquid
LPL
Human recombit protein fragment corresponding to amino acids 28-475 of human LPL produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit