Learn More
Aviva Systems Biology Corp LPP Antibody - N-terminal region (ARP53640_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53640_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LPP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LPP. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-61077