Learn More
Invitrogen™ LPPR5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113795
Description
Immunogen sequence: FTVNVQGFFC HDSAYRKPYP GPEDSSAVPP VLLYSLAAGV PVLVIIVGET For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 83%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The protein encoded by this gene is a type 2 member of the phosphatidic acid phosphatase (PAP) family. All type 2 members of this protein family contain 6 transmembrane regions, and a consensus N-glycosylation site. PAPs convert phosphatidic acid to diacylglycerol, and function in de novo synthesis of glycerolipids as well as in receptor-activated signal transduction mediated by phospholipase D. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq, Jul 2008] Induces filopodia formation and promotes neurite growth in a CDC42-independent manner; impedes neurite growth inhibitory-mediated axonal retraction.
Specifications
LPPR5 | |
Polyclonal | |
Unconjugated | |
PLPPR5 | |
4833424O15Rik; Lipid phosphate phosphatase-related protein type 5; Lppr5; PAP2; Pap2d; phosphatidic acid phosphatase 2d; phosphatidic acid phosphatase type 2; phosphatidic acid phosphatase type 2d; phospholipid phosphatase related 5; phospholipid phosphatase-related protein type 5; Plasticity-related gene 5 protein; plasticity-related protein 5; PLPPR5; PRG5; PRG-5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
163404 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q32ZL2 | |
PLPPR5 | |
Synthetic peptide directed towards the N terminal region of human PAP2D (aa 36-85). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.