Learn More
LRCH2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 248661AP
Catalog No. 50-173-4584
Add to cart
Description
Description
Leucine rich repeat and calponin homology domain containing protein 2 (LRCH2) also named as KIAA1495 is a 765 amino acid protein, which contains one CH domain and nine LRR repeats. The function of LRCH2 is still non-known.
Specifications
Specifications
LRCH2
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3UMG5, Q5VUJ6
dA204F4.4, KIAA1495, LRCH2
Rabbit
IgG
150 μL
-20°C
Primary
210297, 57631
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
LRCH2
Liquid
LRCH2
LRCH2 Fusion Protein Ag21701
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse