Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRCH4 Mouse anti-Human, Clone: OTI1F8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526092

Catalog No. PIMA526092


Add to cart

Description

Description

LRCH4 is a protein that contains leucine-rich repeats (LRR) at its amino terminus and that is known to be involved in ligand binding. The carboxyl terminus may act as a membrane anchor. Identified structural elements suggest that this protein resembles a receptor.This gene encodes a protein that contains leucine-rich repeats (LRR) at its amino terminus and that is known to be involved in ligand binding. The carboxyl terminus may act as a membrane anchor. Identified structural elements suggest that the encoded protein resembles a receptor.
Specifications

Specifications

LRCH4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O75427
Leucine-rich repeats and calponin homology (CH) domain containing 4, FLJ40101, FLJ46315, LRN, LRRN1, LRRN4, PP14183, SAP25
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4034
Immunohistochemistry (Paraffin), Western Blot
OTI1F8
Unconjugated
LRCH4
Liquid
LRCH4
Full length human recombit protein of LRCH4 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit