Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRDD Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157990
Description
LRDD Polyclonal specifically detects LRDD in Human samples. It is validated for Western Blot.Specifications
LRDD | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
leucine-rich repeat and death domain-containing protein, leucine-rich repeats and death domain containing, MGC16925, p53-induced protein with a death domain, PIDDDKFZp434D229 | |
Rabbit | |
Affinity purified | |
RUO | |
55367 | |
Human, Mouse, Rat, Bovine, Canine, Guinea Pig | |
IgG |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9HB75-3 | |
PIDD1 | |
Synthetic peptides corresponding to LRDD(leucine-rich repeats and death domain containing) The peptide sequence was selected from the middle region of LRDD. Peptide sequence RRDPEQVLLQCLPRNKVDATLRRLLERYRGPEPSDTVEMFEGEEFFAAFE. | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction