Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRH-1/NR5A2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $426.00

Specifications

Antigen LRH-1/NR5A2
Concentration 0.5 mg/mL
Dilution Western Blot 2-4ug/ml, Simple Western 1:10
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227204SS
SDP
View Documents
Novus Biologicals
NBP227204SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NBP227204
SDP
View Documents
Novus Biologicals
NBP227204
0.1 mg
Each for $426.00
Only null left
Add to Cart
 
Description

Description

LRH-1/NR5A2 Polyclonal specifically detects LRH-1/NR5A2 in Human, Canine, Chicken, Equine, Feline, Guinea Pig, Opossum, Primate samples. It is validated for Western Blot, Simple Western.
Specifications

Specifications

LRH-1/NR5A2
Western Blot 2-4ug/ml, Simple Western 1:10
Polyclonal
Purified
RUO
Human, Canine, Chicken, Equine, Feline, Guinea Pig, Opossum, Other, Primate
Alpha-1-fetoprotein transcription factor, B1-binding factor, b1-binding factor, hepatocyte transcription factor which activates enhancer IIof hepatitis B virus, B1F, B1F2, fetoprotein-alpha 1 (AFP) transcription factor, FTFLRH1, FTZ-F1, FTZ-F1beta, HB1F, hB1F-2, hB1FCYP7A promoter-binding factor, Hepatocytic transcription factor, Liver receptor homolog 1, liver receptor homolog-1, LRH-1CPF, nuclear receptor NR5A2, nuclear receptor subfamily 5 group A member 2, nuclear receptor subfamily 5, group A, member 2
NR5A2
IgG
Protein A purified
The amino acid sequence used as immunogen for this Lrh-1 antibody is 100% homologous in human, rhesus monkey, orangutan, chimpanzee, new world monkey, panda, dog, horse, african bush elephant, guinea pig, Carolina anole, chicken, opossum, white-cheeked gibbon, platypus, zebra finch, pole-cat, macaque, wild turkey,and monkey, 94% in mole rat, rabbit, sheep, and frog, 88% in pig, ape, and cow, 83% in zebrafish, and 78% homolgous in mouse, and rat.
0.5 mg/mL
Western Blot
Unconjugated
Rabbit
Chromatin Research, Neuroscience
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
2494
A portion of amino acids 300-350 from human Lrh-1 was used as the immunogen for this antibody.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.