Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRIF1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA560734

Catalog No. PIPA560734


Add to cart

Description

Description

Immunogen sequence: SSNYFLTRTV DTSEKGRVTS VGTGNFSSSV SKVQSHGVKI DGLTMQTFAV PPSTQKDSSF IVVNTQSLPV TVKSPVLPSG HHLQIPAHAE VKSVPASSL Highest antigen sequence identity to the following orthologs: Mouse - 62%, Rat - 64%.

The specific function of this protein remains unknown.
Specifications

Specifications

LRIF1
Polyclonal
Unconjugated
LRIF1
Liquid
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
55791
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5T3J3
LRIF1
Recombit protein corresponding to Human LRIF1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Research Use Only