Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRIG2 Mouse anti-Human, Clone: 990332, R&D Systems

Mouse Monoclonal Antibody

$113.85 - $319.00

Specifications

Antigen LRIG2
Clone 990332
Applications Immunocytochemistry, Immunohistochemistry
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB19411100 View Documents R&D Systems
MAB19411100
100 ug Each for $319.00
Add to cart
 
MAB19411SP View Documents R&D Systems
MAB19411SP
25 ug Each for $113.85
Add to cart
 
Description

Description

LRIG2 Monoclonal antibody specifically detects LRIG2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications

Specifications

LRIG2
Immunocytochemistry, Immunohistochemistry
Human
Primary
990332
Mouse
Monoclonal
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit