Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LRIG3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579611
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat testis tissue, HEPG2 whole cell.
May play a role in craniofacial and inner ear morphogenesis during embryonic development. May act within the otic vesicle epithelium to control formation of the lateral semicircular canal in the inner ear, possibly by restricting the expression of NTN1.
Specifications
LRIG3 | |
Polyclonal | |
Unconjugated | |
LRIG3 | |
leucine rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin like domains 3; leucine-rich repeats and immunoglobulin-like domains 3; leucine-rich repeats and immunoglobulin-like domains protein 3; LIG3; LIG-3; LRIG3; RGD1561255; UNQ287/PRO326/PRO335 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
121227, 299830 | |
-20°C | |
Lyophilized |
Western Blot | |
500 μg/mL | |
PBS with 5 mg BSA and 0.05 mg sodium azide | |
Q6UXM1 | |
LRIG3 | |
A synthetic peptide corresponding to a sequence in the middle region of human LRIG3 (428-465aa NAFSQMKKLQQLHLNTSSLLCDCQLKWLPQWVAENNFQ). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction