Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRP-1 Mouse anti-Human, Alexa Fluor™ 594, Clone: A2Mr alpha-2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NB10064808AF594

Catalog No. N1064808A94


Add to cart

Description

Description

LRP-1 Monoclonal antibody specifically detects LRP-1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen).
Specifications

Specifications

LRP-1
A2Mr alpha-2
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen
Conjugated, Purified
LRP1
Purified alpha2 macroglobulin receptor
Protein G purified
Phospho Specific
Primary
4035.0
Human
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
A2MR, Alpha-2-Macroglobulin Receptor, APOER, Apolipoprotein E receptor, APR, CD91, CD91 antigen, EC 1.1.1.94, EC 3.4.21.9, IGFBP3R, low density lipoprotein receptor-related protein 1, LRP, LRP-1, LRP1A, prolow-density lipoprotein receptor-related protein 1, TbetaR-V/LRP-1/IGFBP-3 receptor, TGFBR5, type V tgf-beta receptor
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Recognises an epitope in the extracellular region of the 500kD alpha chain.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit