Learn More
LRP2 Mouse anti-Human, Alexa Fluor 350, Clone: 545606, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB9578U100UG
Catalog No. FAB9578U-100
Due to product restrictions, please to purchase or view availability for this product.
This item is not returnable. View return policy
Description
Description
LRP2 Monoclonal antibody specifically detects LRP2 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
LRP2
545606
Flow Cytometry 0.25-1μg/106 cells
P98164
calcium sensor protein, DBS, DBSlow-density lipoprotein receptor-related protein 2, EC 1.1.2.3, EC 3.4.21.9, Glycoprotein 330, GP330, Heymann nephritis antigen homolog, low density lipoprotein receptor-related protein 2, LRP2, LRP-2, megalin
Mouse myeloma cell line NS0-derived recombinant human Megalin/LRP2 Pro3510-Lys3964 Accession # P98164
Protein A or G purified from ascites
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects human Megalin/LRP2 in direct ELISAs.
Flow Cytometry
Alexa Fluor 350
Supplied 0.2Mg/mL in a saline solution containing BSA and Sodium Azide.
Purified
Mouse
IgG1
100μg
Primary
4036
Human